missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MANT-GTP (2'-(or-3')-O-(N-Methylanthraniloyl) Guanosine 5'-Triphosphate, Trisodium Salt)

Catalog No. M12415
Encompass
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12415 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M12415 Supplier Invitrogen™ Supplier No. M12415
  • ONLINE EXCLUSIVE PRICE
    $594.00
    $487.08 / Each

    Save $106.92

    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Produces nucleotide analogs causing little perturbation of nucleotide-protein interactions

The nucleotide analog MANT GTP is modified on the ribose moiety. The compact mature of the MANT fluorophore and its attachment position results in nucleotide analogs that induce minimal perturbation of nucleotide-protein interactions. Because MANT fluorescence is sensitive to the environment of the fluorophore, nucleotide-protein interactions may be directly detectable. MANT nucleotides are valuable probes of the structure and enzymatic activity of nucleotide-binding proteins.

Drug Discovery and Development, Enzyme and Protein Activity Assays, Phosphatase Activity, Phosphatase Biology, Proteins, Expression, Isolation and Analysis, Target and Lead Identification and Validation

Order Info

Shipping Condition: Wet ice

Specifications

Product Type MANT-GTP
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Shipping Condition Wet Ice
Concentration 5 mM
Label or Dye MANT (N-Methylanthraniloyl)
Quantity 400 μL

For Research Use Only. Not for use in diagnostic procedures.