missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lucifer Yellow CH, Potassium Salt
Water-soluble dye with excitation/emission peaks of 428/536nm
Supplier: Invitrogen™ L1177
Description
- Tool for studying neuronal morphology, because it contains carbohydrazide (CH) group that allows it to be covalently linked to surrounding biomolecules during aldehyde fixation
Cell Analysis, Cell Tracing and Tracking, Neuronal Tracing
Order Info
Shipping Condition: Room temperature
Specifications
| Yellow | |
| Potassium Salt | |
| Store at room temperature and protect from light. | |
| Fluorescence Microscope | |
| 428/536 nm |
| Fluorescence | |
| 25 mg | |
| Room Temperature | |
| Cytoplasm & Cytosol |
For Research Use Only. Not for use in diagnostic procedures.