missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lucifer Yellow CH, Lithium Salt, 25 mg

Catalog No. l453
Encompass
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L453 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L453 Supplier Invitrogen™ Supplier No. L453

Lucifer yellow CH, lithium salt is a water-soluble dye with excitation/emission peaks of 428/536 nm.

Lucifer yellow CH, lithium salt is a water-soluble dye with excitation/emission peaks of 428/536 nm. It is a favorite tool for studying neuronal morphology, because it contains a carbohydrazide (CH) group that allows it to be covalently linked to surrounding biomolecules during aldehyde fixation. The lithium salt form of the lucifer yellow CH is commonly used for microinjection because it has higher solubility than the potassium and ammonium salt forms of lucifer yellow CH.

To make a concentrated stock solution, dissolve in H2O up to 8% (w/v). Store frozen and protected from light.

Order Info

Shipping Condition: Room temperature

Specifications

Color Yellow
Detection Method Fluorescence
Product Type Lithium Salt
Quantity 25 mg
Content And Storage Store at room temperature and protect from light.
Shipping Condition Room Temperature
For Use With (Equipment) Fluorescence Microscope
Sub Cellular Localization Cytoplasm & Cytosol
Excitation/Emission 428/536 nm
Molecular Weight (g/mol) 457.24

For Research Use Only. Not for use in diagnostic procedures.