missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Image-IT™ LIVE Mitochondrial Transition Pore Assay Kit, for microscopy

Catalog No. I35103
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I35103 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I35103 Supplier Invitrogen™ Supplier No. I35103
  • ONLINE EXCLUSIVE PRICE
    $950.00
    $779.00 / Each

    Save $171.00

    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For microscopy

The Image-iT LIVE Mitochondrial Transition Pore Assay Kit provides a proven method [REF PN36389 ,PN32679] of measuring mitochondrial permeability transition pore opening. The kit includes a dye and a quencher that are easily loaded into cells. In healthy cells, the dye, but not the quencher, enters mitochondria. The mitochondria remain brightly fluorescent until mitochondrial pore activation permits quenching of the fluorescence.

Apoptosis, Cell Analysis, Cell Structure, Cell Viability, Proliferation & Function, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Mitochondria, Mitochondria Function, Mitochondrial Function

Order Info

Shipping Condition: Room Temperature

Specifications

Quantity 1 Kit
Format Kit
Product Type Mitochondrial Transition Pore Assay Kit
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Color Orange, Blue, Green
Shipping Condition Room Temperature
Sub Cellular Localization Mitochondria
Product Line Image-iT
For Use With (Equipment) Fluorescence Microscope

For Research Use Only. Not for use in diagnostic procedures.