Learn More
Invitrogen™ Image-IT™ LIVE Green Reactive Oxygen Species Detection Kit, for microscopy DFS Item
For microscopy
Supplier:  Invitrogen™ I36007
Catalog No. I36007
Only null left
Description
Description
  • Image-iT LIVE Green Reactive Oxygen Species (ROS) Detection Kit provides key reagents necessary for detection of ROS in live cells
  • Assay is based on 5-(and-6)-carboxy-2',7'-dichlorodihydrofluorescein diacetate (carboxy-H2DCFDA), reliable fluorogenic marker for ROS in live cells
  • In addition to carboxy-H2DCFDA, kit provides common inducer of ROS production tert-butyl hydroperoxide (TBHP), as positive control and blue-fluorescent, cell-permeant nucleic acid stain Hoechst 33342
Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Nitro-Oxidative Stress, Reactive Oxygen Species
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
1 Kit
Image-iT
ROS Detection Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.