missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HybriWell™ Hybridization Sealing System, 40 mm x 21 mm chamber, 0.15 mm deep

Catalog No. H18210
Encompass
Change view
Click to view available options
Quantity:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H18210 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H18210 Supplier Invitrogen™ Supplier No. H18210
  • ONLINE EXCLUSIVE PRICE
    $520.00
    $426.40 / Each

    Save $93.60

    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Coverslip-seal combination that forms chambers optimized for in situ hybridization work

  • Each disposable incubation chamber seals securely to microscope slide, providing limited-volume chamber for reagent addition
  • Solutions are easily added through dual-access ports without disrupting the specimen
  • Hydrophobic coverslips are nuclease free and will not trap or bind probes, allowing uniform distribution of the reagent over specimen
  • Includes quick-seal tool to secure hydrophobic cover to the microscope slide, as well as nuclease-free adhesive seal tabs to cover the access ports

Cell Analysis, Cellular Imaging, Chromogenic In Situ Hybridization, Fluorescence In Situ Hybridization, In Situ Hybridization (ISH)

Order Info

Shipping Condition: Room temperature

Specifications

Type Hybridization Sealing System
Product Line HybriWell
Depth (Metric) 0.15 mm
Dimensions 40 x 21 mm
Content And Storage Store at room temperature.
Quantity 1 set
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.