missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HybriSlip™ Hybridization Cover, 40 mm x 22 mm, RNase free DFS Item

Designed specifically for in situ hybridization

Supplier:  Invitrogen™ H18201

Catalog No. H18201


Only null left
Explore available promotions

Description

Description

  • Does not require lengthy preparation or blocking procedures to prevent probe trapping or binding
  • Heat resistant and do not curl, even at high temperatures
  • Well-suited for denaturation steps or in situ PCR incubations

Cell Analysis, Cellular Imaging, Chromogenic In Situ Hybridization, Fluorescence In Situ Hybridization, In Situ Hybridization (ISH)

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Hybridization Cover
40 x 22 mm
1 set
HybriSlip
Store at room temperature.
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.