missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Glutathione Ethyl Ester, Biotin Amide (BioGEE) (Glutathiolation Detection Reagent) - Special Packaging

Catalog No. G36000
Encompass
Change view
Click to view available options
Quantity:
10 x 100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G36000 10 x 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G36000 Supplier Invitrogen™ Supplier No. G36000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Glutathiolation detection reagent

Biotinylated glutathione ethyl ester (BioGEE) is a cell-permeant, biotinylated glutathione analog for the detection of glutathiolation. Under conditions of oxidative stress, cells may transiently incorporate glutathione into proteins. Stressed cells incubated in BioGEE will also incorporate this biotinylated glutathione derivative into proteins, facilitating the identification of oxidation-sensitive proteins. Once these cells are fixed and permeabilized, you can detect glutathiolation levels with a fluorescent streptavidin conjugate using either flow cytometry or fluorescence microscopy. Proteins glutathiolated with BioGEE can also be extracted and analyzed by mass spectrometry or by Western blotting methods in conjunction with fluorophore- or enzyme-labeled streptavidin conjugates.

Order Info

Shipping Condition: Room Temperature

Specifications

Quantity 10 x 100 μg
Excitation/Emission 390/405 nm
Product Type Glutathione Reagent

For Research Use Only. Not for use in diagnostic procedures.