missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FocalCheck™ Microspheres, 15 μm, fluorescent dark-red ring stain/green throughout

Catalog No. F7239
Encompass
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F7239 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F7239 Supplier Invitrogen™ Supplier No. F7239

Fluorescent green/orange/dark-red ring stains

The sharp ring stains exhibited by the 15μm FocalCheck fluorescent green/orange/dark-red ring stained microspheres produce a striking visual representation of instrument misalignment or other aberrations making them ideal as reference standards for confocal laser-scanning microscopy. Correct image registration is indicated when the multiple ring images are perfectly coincident in all dimensions.

Cell Analysis, Cell Cycle, Cell Viability, Proliferation and Function, Cellular Imaging, Immunofluorescence (IF)

Specifications

Calibration Type Confocal Microscope Calibration
Content And Storage Store in refrigerator 2°C to 8°C and protect from light.
Color Dark Red, Green
Product Type Microsphere
Format Suspension Beads
Product Line FocalCheck
Quantity 0.5 mL
Shipping Condition Room Temperature
Diameter (Metric) 15 μm

For Research Use Only. Not for use in diagnostic procedures.