missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FocalCheck™ Fluorescence Microscope Test Slide #3, 5 colors, high & low intensities
Description
The FocalCheck fluorescence microscope test slide #3 is specifically designed for calibrating fluorescence microscope systems and evaluating system and filter performance. This slide is useful for basic evaluation of filter performance and as a general practice slide for fluorescence microscopy and digital imaging.
Cell Analysis, Cell Cycle, Cell Viability, Proliferation and Function, Cellular Imaging, Immunofluorescence (IF)
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Calibration Type | Confocal Microscope Calibration, Fluorescence Microscope Calibration |
| Content And Storage | Store at room temperature. |
| Color | Assorted Colors |
| Product Type | Fluorescence Microscope Test Slide |
| Format | Suspension Beads |
| Product Line | FocalCheck |
| Quantity | 1 Ea. |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.