missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FocalCheck™ Fluorescence Microscope Test Slide #2, for spectral imaging systems

Catalog No. F36913
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F36913 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F36913 Supplier Invitrogen™ Supplier No. F36913

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For spectral imaging systems and confocal and fluorescence microscope calibration

The FocalCheck fluorescence microscope test slide #2 is specifically designed for calibrating fluorescence microscope systems and evaluating system and filter performance. This slide provides a robust, reproducible method of evaluating the performance of spectral imaging systems, as well as the ability to discriminate closely overlapping spectra. The slide consists of 6μm-diameter microspheres labeled with a series of spectrally overlapping dyes.

Cell Analysis, Cell Cycle, Cell Viability, Proliferation and Function, Cellular Imaging, Immunofluorescence (IF)

Order Info

Shipping Condition: Room temperature

Specifications

Calibration Type Confocal Microscope Calibration, Fluorescence Microscope Calibration
Content And Storage Store at room temperature.
Color Assorted Colors
Product Type Fluorescence Microscope Test Slide
Format Suspension Beads
For Use With (Equipment) Spectral Imaging System
Product Line FocalCheck
Quantity 1 Ea.
Shipping Condition Room Temperature
Diameter (Metric) 6 μm

For Research Use Only. Not for use in diagnostic procedures.