missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fluorescein - Reference Standard

Catalog No. F1300
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F1300 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F1300 Supplier Invitrogen™ Supplier No. F1300

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use in cell viability, proliferation and function, enzyme and protein activity assays and labeling chemistry

Fluorescein can be used as a reference standard for ether, ester and phosphate substrates derived from this green-fluorescent dye.

  • Label type: Classsic dyes

Cell Analysis, Cell Viability, Proliferation and Function, Enzyme and Protein Activity Assays, Ionic Homeostasis and Signaling, Labeling Chemistry, Labeling QC and Calibration, Protein Assay Controls, Reference Standards and Accessories, Proteins, Expression, Isolation and Analysis, pH Detection

Order Info

Shipping Condition: Room temperature

Specifications

Type Fluorescein Reference Standard
Color Green
Label or Dye Classic Dyes
Content And Storage Store at room temperature and protect from light.
Quantity 1 g
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.