missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fluorescein - NIST-Traceable Standard - Nominal concentration 50μM - Special Packaging

Catalog No. F36915
Encompass
Change view
Click to view available options
Quantity:
5 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F36915 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F36915 Supplier Invitrogen™ Supplier No. F36915
  • ONLINE EXCLUSIVE PRICE
    $688.00
    $564.16 / Each

    Save $123.84

    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Suitable for use in enzyme, protein activity assays and labeling chemistry

The NIST-traceable fluorescein standard is supplied as a calibrated 50μM solution of fluorescein in 100 mM sodium borate buffer, pH 9.5. Under these conditions, fluorescein is completely ionized and is therefore in its most fluorescent form. This standard meets the stringent criteria established by the National Institute of Standards and Technology (NIST) and is directly traceable to Standard Reference Material 1932 (SRM 1932). The NIST-traceable fluorescent standard can be used to assess day-to-day or experiment-to-experiment variation in fluorescence-based instrumentation as well as to determine the Molecules of Equivalent Soluble Fluorophore (MESF) value for an experimental solution.

  • Label type: Classsic dyes

Cell Analysis, Enzyme and Protein Activity Assays, Labeling Chemistry, Labeling QC and Calibration, Protein Assay Controls, Reference Standards and Accessories, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

Specifications

Type Fluorescein Traceable Standard
Label or Dye Classic Dyes
Content And Storage Store in refrigerator 2°C to 8°C and protect from light.
Quantity 5 x 1 mL
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.