missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fluorescein DHPE (N-(Fluorescein-5-Thiocarbamoyl)-1,2-Dihexadecanoyl-sn-Glycero-3-Phosphoethanolamine, Triethylammonium Salt) DFS Item

Excitation/emission maxima ∽496/519nm

Supplier:  Invitrogen™ F362

Catalog No. F362

Only null left
Explore available promotions

Description

Description

The phospholipid, fluorescein DHPE is labeled on the head group with the green-fluorescent pH-sensitive fluorophore, fluorescein

Cell Analysis, Cell Viability, Proliferation & Function, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Ionic Homeostasis & Signaling, pH Detection

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Phospholipids
5 mg
Solid
Invitrogen
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.