missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Epidermal Growth Factor, Fluorescein Conjugate (Fluorescein EGF)

Catalog No. E3478
Encompass
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E3478 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E3478 Supplier Invitrogen™ Supplier No. E3478

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Constructed with the dye directly attached to EGF

Fluorescently labeled EGF has enabled scientists to investigate receptor-membrane interactions, study receptor distribution, calculate rate constants for the interaction of EGF with its receptor, and more.

  • EGF molecule: 53 amino acids, MW=6,045 Da
  • Label (Ex/Em): Fluorescein (approx. 494/518nm)
  • Number of fluorophore molecules on each EGF molecule: 1
  • Fluorescence is typically monitored using a flow cytometer, fluorescence microscope, or fluorimeter

Order Info

Shipping Condition: Wet Ice

Specifications

Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Detection Method Fluorescence
Form Solid
Product Type Growth Factor
Label or Dye Classic Dyes
Quantity 20 μg
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.