missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Epidermal Growth Factor, Biotinylated, complexed to Texas Red™ Streptavidin (Texas Red™ EGF complex)

Catalog No. E3480
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E3480 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E3480 Supplier Invitrogen™ Supplier No. E3480

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Complexes of the dye-streptavidin molecule and biotinylated EGF

Fluorescently labeled EGF has enabled scientists to investigate receptor-membrane interactions, study receptor distribution, calculate rate constants for the interaction of EGF with its receptor, and more.

  • Biotinylated conjugates utilize biotin-XX, which contains long spacer arm designed to enhance probe affinity for EGF receptor
  • EGF molecule: 53 amino acids, MW=6,045 Da
  • Number of fluorophore molecules on each EGF molecule: 1
  • Fluorescence is typically monitored using a flow cytometer, fluorescence microscope, or fluorimeter

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Wet Ice

Specifications

Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Cell Type Mammalian Cells
Detection Method Fluorescence
Product Type Growth Factor
Label or Dye Classic Dyes
Product Line Texas Red
Quantity 100 μg
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.