missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dihydrorhodamine 123

Catalog No. D632
Encompass
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D632 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D632 Supplier Invitrogen™ Supplier No. D632
  • ONLINE EXCLUSIVE PRICE
    $490.00
    $401.80 / Each

    Save $88.20

    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

missing translation for 'validMainframeMsg'

Uncharged and nonfluorescent reactive oxygen species indicator

Dihydrorhodamine 123 can passively diffuse across membranes where it is oxidized to cationic rhodamine 123 which localizes in the mitochondria and exhibits green fluorescence.

Concentration: 5mM

Applications:

Apoptosis, Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Free Radical Detection, Nitro-Oxidative Stress, Reactive Oxygen Species

Specifications

Quantity 10 mg
Shipping Condition Room Temperature
I am using the reactive oxygen species detection dye, Dihydrorhodamine 123, and when I put the dye in a solution of water and illuminate with UV light, I see a quick change in signal, even without cells. What is happening and how can I stop this?

You cannot stop it. Any time you illuminate a dye, including this one, photodegradation of the dye will lead to the production of singlet oxygen and free radicals. This in turn causes many ROS indicators, like this one, to become oxidized. If you are testing ROS using UV illumination, all you can do is use this as a baseline and subtract the signal from the sample with cells. The best you can do is reduce the illumination intensity (though this will also reduce the specific signal emission).

For Research Use Only. Not for use in diagnostic procedures.