missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dihydroethidium (Hydroethidine)

Free radical sensor

$494.00 - $626.00


Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
D11347
Encompass
View Documents Promotion Details
Invitrogen™
D11347
10 x 1 mg
Each for $626.00
Only null left
 
D1168
Encompass
View Documents Promotion Details
Invitrogen™
D1168
25 mg
Each for $494.00
Only null left
 
Description

Description

  • Superoxide indicator dihydroethdium, also called hydroethidine, exhibits blue-fluorescence in cytosol until oxidized, where it intercalates within cell's DNA, staining its nucleus a bright fluorescent red
  • Excitation Emission: 518/606nm

Cat. No. D23107 packaged 5mM solution stabilized in DMSO.

Apoptosis, Cell Analysis, Cell Structure, Cell Tracing and Tracking, Cell Viability, Proliferation and Function, Free Radical Detection, Nitro-Oxidative Stress, Nucleus, Nucleoli and Nuclear Envelope, Organelle Tracing, Reactive Oxygen Species

Order Info

Shipping Condition: Room Temperature

Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.