missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Di(Acetoxymethyl Ester) (6-Carboxy-2',7'-Dichlorodihydrofluorescein Diacetate)

Catalog No. C2938
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2938 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2938 Supplier Invitrogen™ Supplier No. C2938

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemically reduced, acetylated form of fluorescein used as indicator for reactive oxygen species in cells.

6-carboxy-2',7'-dichlorodihydrofluorescein diacetate is a chemically reduced, acetylated form of fluorescein used as an indicator for reactive oxygen species (ROS) in cells. This nonfluorescent molecule is readily converted to a green-fluorescent form when the acetate groups are removed by intracellular esterases and oxidation (by the activity of ROS) occurs within the cell. We offer the carboxylated H2DCFDA analog (carboxy-H2DCFDA, (Cat. No. C400)), which has two negative charges at physiological pH, and its di(acetoxymethyl ester) (Cat. No. C2938). Upon cleavage of the acetate and ester groups by intracellular esterases and oxidation, both analogs form carboxydichlorofluorescein (Cat. No. C368), with additional negative charges that impede its leakage out of the cell.

ROS Indicator Specifications:

  • Ex/Em: ∼492–495/517–527 nm
  • Product is air sensitive and should be stored under dry argon or nitrogen
  • Product may be dissolved in DMSO, DMF, or ethanol for use
  • Indicator is cell permeant (cell loading protocols are available in the literature)
  • Fluorescence can be monitored using a flow cytometer, fluorometer, microplate reader, or fluorescence microscope, using excitation sources and filters appropriate for fluorescein

    Find More ROS Indicators

    We offer an assortment of Molecular Probes™ products for the generation of reactive oxygen species (ROS), including singlet oxygen, superoxide, hydroxyl radical and various peroxide and hydroperoxides, as well as for their fluorometric detection in solution. Review Generating and Detecting Reactive Oxygen Species—Section 18.2 in the Molecular Probes™ Handbook for more information on these products.

  • Order Info

    Shipping Condition: Room Temperature

    Specifications

    Quantity 5 mg
    Detection Method Fluorescence
    Product Type ROS Indicator

    For Research Use. Not for human or animal therapeutic or diagnostic use.