missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dextran, Cascade Blue™, 3000 MW, Anionic, Lysine Fixable

Catalog No. d7132
Encompass
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D7132 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D7132 Supplier Invitrogen™ Supplier No. D7132

Rigorous methods for removing as much unconjugated dye as practical, followed by assay of dextran conjugates by thin-layer chromatography to help ensure absence of low molecular weight contaminants

Labeled dextrans are hydrophilic polysaccharides most commonly used in microscopy studies to monitor cell division, track the movement of live cells, and to report the hydrodynamic properties of the cytoplasmic matrix. The labeled dextran is commonly introduced into the cells via microinjection. Dextrans are hydrophilic polysaccharides characterized by their moderate-to-high molecular weight, good water solubility, and low toxicity. They also generally exhibit low immunogeniticy. Dextrans are biologically inert due to their uncommon poly-(α-D-1,6-glucose) linkages, which render them resistant to cleavage by most endogenous cellular glycosidases.

  • Label (Ex/Em): Cascade Blue (400/420)
  • Size: 3000 MW
  • Charge: Anionic
  • Fixable: Fixable via Lysine
  • Succinimidyl coupling of dyes to dextran molecule, which, in most cases, results in a neutral or anionic dextran
  • Reaction used to produce Rhodamine Green™ and Alexa Fluor™ 488 dextrans results in final product being neutral, anionic, or cationic
  • Common uses include neuronal tracing (anterograde and retrograde) in live cells
  • Cell lineage tracing in live cells
  • Neuroanatomical tracing
  • Examines intercellular communications (e.g., in gap junctions, during wound healing, and during embryonic development)
  • Investigates vascular permeability and blood-brain barrier integrity
  • Tracks endocytosis
  • Monitors acidification (some dextran-dye conjugates are pH-sensitive)
  • Studies hydrodynamic properties of cytoplasmic matrix
    • Cascade Blue dextrans are intrinsically anionic
    • To produce more highly anionic dextrans, proprietary procedure adds negatively charged groups to dextran carriers; these products are designated polyanionic dextrans
    • Some applications require that dextran tracer be treated with formaldehyde or glutaraldehyde for subsequent analysis: For these applications, lysine-fixable dextran conjugates of fluorophores or biotin
    • Dextrans have covalently bound lysine residues that permit dextran tracers to be conjugated to surrounding biomolecules by aldehyde-mediated fixation for subsequent detection by immunohistochemical and ultrastructural techniques

    Cell Analysis, Cell Tracing and Tracking, General Cell Tracing, Neuronal Tracing

Order Info

Shipping Condition: Room temperature

Specifications

Product Type Dextran
Quantity 10 mg
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label or Dye Other Label(s) or Dye(s)
Shipping Condition Room Temperature
Excitation/Emission 400/420 nm
Product Line CASCADE BLUE

For Research Use Only. Not for use in diagnostic procedures.