missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dextran, Amino, 10,000 MW

Catalog No. D1860
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1860 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D1860 Supplier Invitrogen™ Supplier No. D1860
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rigorous methods for removing as much unconjugated dye as practical, followed by an assay of dextran conjugates by thin-layer chromatography to help ensure the absence of low molecular weight contaminants

Labeled dextrans are hydrophilic polysaccharides most commonly used in microscopy studies to monitor cell division, track the movement of live cells, and to report the hydrodynamic properties of the cytoplasmic matrix. The labeled dextran is commonly introduced into the cells via microinjection. Dextrans are hydrophilic polysaccharides characterized by their moderate-to-high molecular weight, good water solubility, and low toxicity. They also generally exhibit low immunogeniticy. Dextrans are biologically inert due to their uncommon poly-(α-D-1,6-glucose) linkages, which render them resistant to cleavage by most endogenous cellular glycosidases.

  • Succinimidyl coupling of dyes to dextran molecule, which, in most cases, results in neutral or anionic dextran
  • Suitable for neuronal tracing (anterograde and retrograde) in live cells
  • Cell lineage tracing in live cells
  • Examining intercellular communications (e.g., in gap junctions, during wound healing, and during embryonic development)
  • Investigating vascular permeability and blood-brain barrier integrity
  • Monitoring acidification (some dextran-dye conjugates are pH-sensitive)
  • Studying hydrodynamic properties of cytoplasmic matrix

Cell Analysis, Cell Tracing and Tracking, General Cell Tracing, Neuronal Tracing

Order Info

Shipping Condition: Room temperature

Specifications

Product Type Dextran
Quantity 1 g
Content And Storage Store at room temperature.
Label or Dye Unlabeled
Shipping Condition Room Temperature
Product Line Invitrogen

For Research Use Only. Not for use in diagnostic procedures.