missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DAF-FM Diacetate (4-Amino-5-Methylamino-2',7'-Difluorofluorescein Diacetate)

Catalog No. p-4924916
Encompass
Change view
Click to view available options
Quantity:
10 x 50 μg
1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23844 10 x 50 μg
D23842 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23844 Supplier Invitrogen™ Supplier No. D23844

For detection and quantification of low concentrations of nitric oxide (NO).

It is essentially nonfluorescent until it reacts with NO to form a fluorescent benzotriazole. DAF-FM fluorescence can be detected by any instrument that can detect fluorescein, including flow cytometers, microscopes, fluorescent microplate readers and fluorometers.

  • Ex/Em of DAF-FM: approx. 495/515nm
  • Lyophilized product should be dissolved using DMSO and then added to an aqueous buffer to create a working solution
  • DAF-FM diacetate is cell permeant and passively diffuses across cellular membranes; once inside the cell, it is converted to cell-impermeant for
  • Buffers containing bovine serum albumin (BSA) and phenol red may affect fluorescence and should be used with caution
  • Fluorescence quantum yield of DAF-FM is approx. 0.005, but increases about 160-fold, to ∼0.81, after reacting with NO
  • Assessment of NO production in transaldolase-deficient lymphoblasts by flow cytometry
  • Detection of NO accumulation in embryonic cortical neurons following neurotrophin stimulation
  • in vivo imaging of NO in zebrafish
  • Intravital microscopic detection of NO generation associated with angiogenesis in mice
  • Quantitation of ATP-induced NO release in rabbit platelets
  • Spectra of the NO adduct of DAF-FM are independent of pH above pH 5.5
  • NO adduct of DAF-FM is significantly more photostable than that of DAF-2, which means additional time for image capture
  • DAF-FM is a more sensitive reagent for NO than is DAF-2 (NO detection limit for DAF-FM approx. 3nM versus approx. 5nM for DAF-2)

Refer to the Molecular Probes™ Handbook for additional product information.

Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Nitric Oxide Research, Nitro-Oxidative Stress

Order Info

Shipping Condition: Room Temperature

Specifications

Quantity 10 x 50 μg
Product Type NO Indicator

For Research Use Only. Not for use in diagnostic procedures.