missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CultureWell™ Multiwell Chambered Coverslip CWCS 8R-1.0, 24 mm x 50 mm coverslips, eight 1 mm-deep wells per coverslip DFS Item

Convenient for preparing cultured cells for staining and imaging

Supplier:  Invitrogen™ C24779

Catalog No. C24779

Only null left
Explore available promotions

Description

Description

The CultureWell cell culture chambered coverslips use medical-grade silicone gaskets with standard optical-quality coverslips and are provided sterile and ready to use. Preassembled CultureWell systems are also available.

Cell Analysis, Cell Culture, Cellular Imaging, IHC Sample Preparation, IHC Staining Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Sample Preparation, Immunofluorescence Staining Detection, Immunohistochemistry (IHC), Mammalian Cell Culture

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Slide Coverslip
1 mm
1 mm
Room Temperature
Sterile
8
Store at room temperature.
6 mm
50 mm x 24 mm
1 set
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.