missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CoverWell™ Perfusion Chamber Gasket, 8 chambers, 9 mm diameter, 0.5 mm deep

Catalog No. C18139
Encompass
Change view
Click to view available options
Quantity:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C18139 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C18139 Supplier Invitrogen™ Supplier No. C18139
  • ONLINE EXCLUSIVE PRICE
    $280.00
    $229.60 / Each

    Save $50.40

    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed to facilitate live-cell imaging and manipulation

  • CoverWell perfusion chamber gaskets form watertight “press-to-seal” chambers with dual-access ports so reagents can be quickly added and removed
  • Available in single- or multi-well configurations, these gaskets allow multiple experiments to be performed on single microscope slide or coverslip
  • Heat-resistant gaskets can be sterilized and used for direct culturing of cells and tissues

Applications:

Cell Analysis, Cellular Imaging, IHC Sample Preparation, IHC Staining & Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Staining & Detection, Immunohistochemistry (IHC)

Specifications

Type Perfusion Chamber Gasket
Color Red
Thickness (Metric) 0.5 mm
Content And Storage Store at room temperature.
Depth (Metric) 0.5 mm
Diameter (Metric) 9 mm
Shipping Condition Room Temperature
Product Line CoverWell
Sterility Sterile
Quantity 1 set

For Research Use Only. Not for use in diagnostic procedures.