Learn More
Invitrogen™ CoverWell™ Incubation Chamber Gasket
Designed for immunocytochemistry and in situ hybridization
Supplier: Invitrogen™ C18155
Description
These heat resistant, autoclavable and nuclease free chamber gaskets are designed for use with incubation chambers. The chambers improve staining uniformity and sensitivity by enclosing a large sample area while minimizing the reagent volume required. The watertight seal holds reactants in place and prevents evaporation even during prolonged heating and is easily removed for multiple-step procedures.
Applications:
Cell Analysis, Cellular Imaging, IHC Sample Preparation, IHC Staining and Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Sample Preparation, Immunofluorescence Staining and Detection, Immunohistochemistry (IHC)
Specifications
| Incubation Chamber Gasket | |
| 0.2 mm | |
| 0.2 mm | |
| Room Temperature | |
| CoverWell |
| Red | |
| Store at room temperature. | |
| 13 mm | |
| 25 gaskets | |
| 25 gaskets |
For Research Use Only. Not for use in diagnostic procedures.