missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CoverWell™ Incubation Chamber Gasket

Catalog No. C18151
Encompass
Change view
Click to view available options
Quantity:
50 gaskets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C18151 50 gaskets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C18151 Supplier Invitrogen™ Supplier No. C18151

Designed for immunocytochemistry and in situ hybridization

These heat resistant, autoclavable and nuclease free chamber gaskets are designed for use with incubation chambers. The chambers improve staining uniformity and sensitivity by enclosing a large sample area while minimizing the reagent volume required. The watertight seal holds reactants in place and prevents evaporation even during prolonged heating and is easily removed for multiple-step procedures.

Applications:

Cell Analysis, Cellular Imaging, IHC Sample Preparation, IHC Staining and Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Sample Preparation, Immunofluorescence Staining and Detection, Immunohistochemistry (IHC)

Specifications

Type Incubation Chamber Gasket
Color Red
Thickness (Metric) 0.5 mm
Content And Storage Store at room temperature.
Depth (Metric) 0.5 mm
Shipping Condition Room Temperature
Dimensions (LxW) 40 mm x 22 mm
Product Line CoverWell
Quantity 50 gaskets

For Research Use Only. Not for use in diagnostic procedures.