missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Click-IT™ Tetramethylrhodamine (TAMRA) Protein Analysis Detection Kit
GREENER_CHOICE

Catalog No. c33370
Encompass
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 Kit
Catalog No. Quantity
C33370 1 Kit
1 options

Catalog No. C33370

Supplier: Invitrogen™ C33370

Only null left

Supports differential analysis of glycoprotein subclasses, total proteins and total glycoproteins in the same gel

The Click-iT Tetramethylrhodamine (TAMRA) Glycoprotein Detection Kit provides the second part of the simple and robust two-step technique to identify and characterize glycoproteins by one- or two-dimensional gel electrophoresis. In step two, after the incorporation of the azide handle into protein glycan structures with either a Click-iT metabolic labeling reagent or the Click-iT Enzymatic Labeling system, the azide-modified glycoproteins are detected via the chemoselective ligation or click reaction between an azide and an alkyne. Gels with TAMRA labeled glycoproteins can be subsequently stained with the Multiplexed Proteomics™ technologies, SYPRO™ Ruby total protein stain and PRO-Q™ Emerald glycoprotein stain for the differential analysis of glycoprotein subclasses, total proteins and total glycoproteins in the same gel. Click-iT modified glycoproteins are also compatible with downstream LC-MS/MS and MALDI MS analysis for identification.

  • Less hazardous

Cell Analysis, Click Chemistries, Glycoprotein Analysis with Click Chemistry, Labeling Chemistry, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

Specifications

Green Features Less hazardous
Quantity 1 Kit
Product Type TAMRA Protein Analysis Detection Kit
Content And Storage Store in freezer -5°C to -30°C.
Detection Method Fluorescence
Shipping Condition Room Temperature
Label or Dye TAMRA™ Isomers, TMR (Tetramethylrhodamine)
Product Line Click-iT
Labeling Target Proteins (Glycoproteins)

For Research Use Only. Not for use in diagnostic procedures.