missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cell Culture Contamination Detection Kit

Catalog No. C7028
Encompass
Change view
Click to view available options
Quantity:
200 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C7028 200 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C7028 Supplier Invitrogen™ Supplier No. C7028
  • ONLINE EXCLUSIVE PRICE
    $485.00
    $397.70 / Each

    Save $87.30

    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Detects and identifies contaminants

The Cell Culture Contamination Detection Kit is a simple and effective procedure for screening tissue cultures for contamination by microorganisms. The kit not only detects contaminants, but it also identifies the contaminant type. Three fluorescent dyes distinctly stain yeast (and other fungi), gram-positive and gram negative bacteria by slide preparation.

Bioproduction, Cell Analysis, Cell Culture, Cell Tracing and Tracking, Clinical, Contaminant and Impurity QC Testing, Infectious Disease Diagnostics, Mammalian Cell Culture, Microbial Tracking, Molecular Diagnostics, Mycoplasma Detection

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Detection Method Fluorescence
Form Liquid
Product Type Cell Culture Contamination Detection
For Use With Fluorescence Microscope
Quantity 200 Assays
Shipping Condition Room Temperature
What does the Cell Culture Contamination Detection kit screen for?

This kit can screen tissue cultures for contamination by microorganisms. The kit not only serves to detect the contaminants, it also identifies the contaminant type. Three fluorescent dyes distinctly stain yeast (and other fungi), gram-positive and gram-negative bacteria in slide preparations. The kit provides results within one hour, and, by staining the microorganisms with different fluorescent colors, it helps a scientist choose an appropriate course of action.

Can the Cell Culture Contamination Detection Kit (Cat. No. C7028) detect mycoplasma?

Yes, the Cell Culture Contamination Detection Kit will detect/stain mycoplasma. The SYTO-9 cell-permeant dye will stain every cell, including the mammalian cells (if present). Mycoplasma will be seen as small contaminating cells in the medium. It may be challenging to identify Mycoplasma from other possible contaminating species, if present.

We also offer the MycoFluor Mycoplasma Detection Kit (Cat. No. M7006) for mycoplasma detection in lab cultures.

For Research Use Only. Not for use in diagnostic procedures.