missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BrdUTP (5-Bromo-2'-Deoxyuridine 5'-Triphosphate), 10 mM in TE buffer

Catalog No. B21550
Encompass
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21550 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B21550 Supplier Invitrogen™ Supplier No. B21550
  • ONLINE EXCLUSIVE PRICE
    $310.00
    $254.20 / Each

    Save $55.80

    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Label apoptotic cells

BrdUTP can be used in conjunction with tetrminal deoxynucleotidyl transferase (TdT) to label apoptotic cells in the TUNEL assay. BrdUTP can be detected with anti-BrdU antibodies.

Apoptosis, Cell Analysis, Cell Proliferation, Cell Viability, Proliferation and Function, Chromatin Biology, DNA Fragmentation, RNAi, Epigenetics and Non-Coding RNA Research

Order Info

Shipping Condition: Wet ice

Specifications

Product Type BrdUTP
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Shipping Condition Wet Ice
Concentration 10 mM
Label or Dye Other Label(s) or Dye(s)
Quantity 25 μL
Can dU-containing DNA produced by amplification with dUTP be cut by restriction enzymes?

You will need to confirm with the manufacturer of the restriction enzyme you are using to find out whether or not it can recognize dUTP-containing DNA.

For Research Use Only. Not for use in diagnostic procedures.