missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BrainStain™ Imaging Kit

Catalog No. B34650
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34650 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B34650 Supplier Invitrogen™ Supplier No. B34650
  • ONLINE EXCLUSIVE PRICE
    $860.00
    $705.20 / Each

    Save $154.80

    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Compatible with standard histochemical methods such as immunohistochemistry

The BrainStain Imaging Kit enables three-color combinatorial labeling of myelin, neurons, and nuclei in brain cryosections in a single 20-minute staining step. The kit contains novel stains that can be used together in one staining solution or separately and replaces traditional methods that can take one to three days.

Cell Analysis, Cell Tracing & Tracking, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Neuronal Tracing

Order Info

Shipping Condition: Room Temperature

Specifications

Color Green, Red, Blue
Detection Method Fluorescence
Product Type Imaging Kit
Quantity 1 kit
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label or Dye NeuroTrace™ 530⁄615, DAPI, FluoroMyelin™ Green
Shipping Condition Room Temperature
For Use With (Equipment) Fluorescence Microscope
Sub Cellular Localization Nucleus, Myelin, Nissl Bodies
Product Line BrainStain
What dyes are included in the BrainStain Imaging Kit?

The BrainStain Imaging Kit enables three-color staining of myelin, neurons, and nuclei in brain cryosections in a single 20-minute staining step. It includes FluoroMyelin Green dye, NeuroTrace 530/615 red fluorescent Nissl stain, and DAPI for staining nuclei.

I want to stain brain cryosections. Do you have any kits for this purpose?

We offer the BrainStain Imaging Kit (Cat. No. B34650) for staining brain cryosections. This kit enables three-color labeling of myelin, neurons, and nuclei in brain cryosections in a single 20-minute staining step.

For Research Use Only. Not for use in diagnostic procedures.