Learn More
Invitrogen™ BODIPY™ FL C5-Ganglioside GM1
Useful in cellular imaging
Supplier: Invitrogen™ B13950
Description
Colocalization of fluorescent cholera toxin B conjugates and the green-fluorescent BODIPY FL C5-ganglioside GM1 observed by fluorescence microscopy provides a direct indication of the association of these molecules in lipid rafts.
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
- Environmental benefits include:
- Renewable Energy
Specifications
Sphingolipids | |
25 ÎĽg | |
Solid | |
BODIPY |
For Research Use Only. Not for use in diagnostic procedures.