missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BODIPY™ FL C5 (4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Pentanoic Acid)

Catalog No. D3834
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3834 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D3834 Supplier Invitrogen™ Supplier No. D3834

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fluorescent in both aqueous and lipid environments

The green fluorescent fatty acid, BODIPY FL C5 can be used as a synthetic precursor to a wide variety of fluorescent phospholipids. Unlike fluorophores like pyrene, DPH and NBD, BODIPY dyes are relatively environment insensitive.

Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection

Order Info

Shipping Condition: Room Temperature

Specifications

Chemical Name or Material Fatty Acids
Quantity 1 mg
Physical Form Solid
Product Line BODIPY

For Research Use Only. Not for use in diagnostic procedures.