missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BODIPY™ FL C16 (4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Hexadecanoic Acid)

Catalog No. d3821
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3821 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D3821 Supplier Invitrogen™ Supplier No. D3821
Only null left

The green fluorescent fatty acid, BODIPY™ FL C16 can be used as a synthetic precursor to a wide variety of fluorescent phospholipids.

The green fluorescent fatty acid, BODIPY™ FL C16 can be used as a synthetic precursor to a wide variety of fluorescent phospholipids. Unlike fluorophores like pyrene, DPH and NBD, BODIPY™ dyes are relatively environment insensitive and is fluorescent in both aqueous and lipid environments.

We recommend dissolving in high-quality anhydrous DMSO to attain a stock concentration of 1 to 10 mM.

Order Info

Shipping Condition: Room Temperature

Specifications

Chemical Name or Material Fatty Acids
Quantity 1 mg
Physical Form Solid
Product Line BODIPY
Excitation/Emission 505/512 nm

For Research Use Only. Not for use in diagnostic procedures.