Learn More
Invitrogen™ BODIPY™ FL C12-Sphingomyelin (N-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Dodecanoyl) Sphingosyl Phosphocholine)
Green-fluorescent sphingomyelin
Supplier: Invitrogen™ D7711
Description
BODIPY FL C12-sphingomyelin can be used to study sphingolipid transport and signaling pathway in cells.
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Sphingolipids | |
250 ÎĽL | |
BODIPY |
For Research Use Only. Not for use in diagnostic procedures.