missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BODIPY™ 665/676 (Lipid Peroxidation Sensor)

Catalog No. b3932
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3932 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B3932 Supplier Invitrogen™ Supplier No. B3932

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BODIPY 665/676 is a lipophilic probe used to detect lipid peroxidation. It exhibits a change in fluorescence after interaction with peroxyl radicals within cells.

BODIPY 665/676 is a lipophilic probe used to detect lipid peroxidation. It exhibits a change in fluorescence after interaction with peroxyl radicals within cells. We recommend dissolving BODIPY 665/676 in high-quality anhydrous DMSO to attain a stock concentration of 1 to 10 mM.

Lipid peroxidation is the oxidative degradation of cellular lipids by free radicals from reactive oxygen species, which can degrade lipids containing carbon-carbon double bonds such as phospholipids and polyunsaturated fatty acids. Oxidation of lipids leads to the generation of lipid peroxides and can cause damage to cell membranes, resulting in changes to signal transduction pathways and eventually leading to cell death. Lipid peroxidation is involved in apoptosis and one of the main contributors to ferroptosis, an iron-dependent, non-apoptotic form of cell death. Oxidative stress from lipid peroxidation plays a role in aging, as well as pathologies such as cancer, atherosclerosis, and neurodegenerative diseases.

Order Info

Shipping Condition: Room Temperature

Specifications

Color Far-red
Quantity 5 mg
For Use With (Application) Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Nitro-Oxidative Stress, Reactive Oxygen Species
Shipping Condition Room Temperature
Target Function Lipid peroxidation
Detection Method Fluorescence
Excitation/Emission 665/676 nm
Indicator ROS
Product Line BODIPY
Product Type Lipid Sensor

For Research Use Only. Not for use in diagnostic procedures.