missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BODIPY™ 581/591 C11 (Lipid Peroxidation Sensor)

Catalog No. D3861
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3861 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D3861 Supplier Invitrogen™ Supplier No. D3861
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Lipid peroxidation sensor

BODIPY™ 581/591 C11 undecanoic acid (Lipid Peroxidation Sensor) is a sensitive fluorescent probe for lipid peroxidation assays that localizes to membranes in live cells. Oxidation of the polyunsaturated butadienyl portion of this fatty acid analog in live cells results in a shift of the fluorescence emission peak from red (∼590 nm) to green (∼510 nm), allowing ratiometric analysis of lipid peroxidation using fluorescence microscopy, high-content analysis, and flow cytometry. In the reduced state, the excitation and emission maxima of BODIPY 581/591 C11 is 581/591 nm; after oxidation, the probe shifts the excitation and emission to 488/510 nm.

Lipid peroxidation is the oxidative degradation of cellular lipids by free radicals from reactive oxygen species, which can degrade lipids containing carbon-carbon double bonds such as phospholipids and polyunsaturated fatty acids. Oxidation of lipids leads to the generation of lipid peroxides and can cause damage to cell membranes, resulting in changes to signal transduction pathways and eventually leading to cell death. Lipid peroxidation is involved in apoptosis and one of the main contributors to ferroptosis, an iron-dependent, non-apoptotic form of cell death. Oxidative stress from lipid peroxidation plays a role in aging, as well as pathologies such as cancer, atherosclerosis, and neurodegenerative diseases.

The BODIPY 581/591 C11 reagent provides a sensitive method to measure lipid peroxidation that occurs in these biological processes and pathological conditions. Because it shifts its fluorescence emission upon oxidation and can be measured through ratiometric fluorescence analysis at two wavelengths, measurement with the BODIPY 581/591 C11 reagent can minimize variations in fluorescence intensity due to factors such as indicator concentration, excitation intensity, and photobleaching. The BODIPY 581/591 C11 reagent can also be used for fluorometric lipid peroxidation assays of antioxidant efficacy in plasma and lipid vesicles.

The BODIPY™ 581/591 C11 (Lipid Peroxidation Sensor) reagent is also available in the Image-iT™ Lipid Peroxidation Kit (Cat. No. C10445) which includes cumene hydroperoxide as a positive control to induce lipid peroxidation in live cells.

We recommend dissolving in high-quality anhydrous DMSO to attain a stock concentration of 1 to 10 mM.

Order Info

Shipping Condition: Room Temperature

Specifications

Quantity 1 mg
Detection Method Fluorescence
Excitation/Emission 581/591 nm
Indicator ROS
Product Line BODIPY
Product Type Lipid Sensor
What solvent should be used to reconstitute/dissolve BODIPY 581/591 C11 (Lipid Peroxidation Sensor) (Cat. No. D3861)?

For BODIPY 581/591 C11 (Cat No. D3861), we recommend that you dissolve this in dimethyl sulfoxide (DMSO). You can prepare a stock solution in any concentration between 1 and 5 mg/mL. Please note that the BODIPY/DMSO stock solution must be stored at -20 degrees C, desiccated and protected from light.

For Research Use Only. Not for use in diagnostic procedures.