missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BODIPY™ 558/568 C12 (4,4-Difluoro-5-(2-Thienyl)-4-Bora-3a,4a-Diaza-s-Indacene-3-Dodecanoic Acid)

Catalog No. D3835
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3835 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D3835 Supplier Invitrogen™ Supplier No. D3835

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The orange-red fluorescent fatty acid, BODIPY™ 558/568 C12 can be used as a synthetic precursor to a wide variety of fluorescent phospholipids.

The orange-red fluorescent fatty acid, BODIPY™ 558/568 C12 can be used as a synthetic precursor to a wide variety of fluorescent phospholipids. Unlike fluorophores like pyrene, DPH and NBD, BODIPY™ dyes are relatively environment insensitive and is fluorescent in both aqueous and lipid environments.

We recommend dissolving in high-quality anhydrous DMSO to attain a stock concentration up to 1 mM.

Order Info

Shipping Condition: Room Temperature

Specifications

Chemical Name or Material Fatty Acids
Quantity 1 mg
Physical Form Solid
Product Line BODIPY
Excitation/Emission 559/568 nm

For Research Use Only. Not for use in diagnostic procedures.