Learn More
Invitrogen™ BODIPY™ 500/510 C1, C12 (4,4-Difluoro-5-Methyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Dodecanoic Acid)
Forms excimers and exhibits red-shifted fluorescence emission when incorporated into living cells
Supplier: Invitrogen™ D3823
Description
C1-BODIPY 500/510 C12 is a fluorescent fatty acid analog that has been used for lipid trafficking studies and as a general membrane probe.
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
Specifications
| Fatty Acids | |
| 1 mg | |
| Solid | |
| BODIPY |
For Research Use Only. Not for use in diagnostic procedures.