missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ β-BODIPY™ FL C12-HPC (2-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Dodecanoyl)-1-Hexadecanoyl-sn-Glycero-3-Phosphocholine)

Catalog No. D3792
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3792 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D3792 Supplier Invitrogen™ Supplier No. D3792

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Effective lipid trafficking tracer and general-purpose membrane probes

The intensely fluorescent BODIPY (4,4-difluoro-3a,4a-diaza-
s-indacene) fluorophore is intrinsically lipophilic, unlike most other long-wavelength dyes. Consequently, probes incorporating this fluorophore are more likely to mimic the properties of natural lipids. BODIPY™ fatty acid analogs like BODIPY 500/510 dodecanoic acid (D-3823) and BODIPY FL dodecanoic acid (D-3822) undergo native-like transport and metabolism in cells.

Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection

Order Info

Shipping Condition: Room Temperature

Specifications

Chemical Name or Material Phospholipids
Quantity 100 μg
Physical Form Solid
Product Line BODIPY

For Research Use Only. Not for use in diagnostic procedures.