missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Attofluor™ Cell Chamber, for microscopy

Catalog No. A7816
Encompass
Change view
Click to view available options
Quantity:
3 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A7816 3 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A7816 Supplier Invitrogen™ Supplier No. A7816
  • ONLINE EXCLUSIVE PRICE
    $778.00
    $637.96 / Each

    Save $140.04

    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Durable and practical coverslip holder designed for viewing live cell specimens on upright or inverted microscopes

  • Because chamber is constructed from surgical stainless steel and is autoclavable, cells can be grown directly on coverslips within the unit
  • O-ring seal design prevents samples from being contaminated by oil or leakage of media from coverslips
  • Chamber accepts 25mm round coverslips and mounts in standard 35mm diameter stage holder, and 0.5mm base dimension allows clearance for objective when focusing

Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Sample Preparation, Immunofluorescence Staining and Detection

Order Info

Shipping Condition: Room temperature

Specifications

Type Imaging System Cell Chamber
Product Line Attofluor
Diameter (Metric) 35 mm
Content And Storage Store at room temperature.
Quantity 3 Pack
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.