missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ANTS (8-Aminonaphthalene-1,3,6-Trisulfonic Acid, Disodium Salt)

Catalog No. a350
Encompass
Change view
Click to view available options
Quantity:
1 g
Catalog No. Quantity
A350 1 g
1 options

Catalog No. A350

Supplier: Invitrogen™ A350

Only null left

Polyanionic dye often used in combination with the cationic quencher DPX for membrane fusion or permeability assays, including complement-mediated immune lysis

  • The unusually high Stokes shift of ANTS in water (>150nm) separates this from much of the autofluorescence of biological samples
  • ANTS has also been employed as a neuronal tracer

Order Info

Shipping Condition: Room temperature

Specifications

Quantity 1 g
Shipping Condition Room Temperature
For Use With (Equipment) Fluorescence Microscope
Sub Cellular Localization Cell Membranes & Lipids
Detection Method Fluorescence
Molecular Formula C10H7NNa2O9S3
Physical Form Solid
Color Green
Product Type Cell Tracer

For Research Use Only. Not for use in diagnostic procedures.