missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Albumin from Bovine Serum (BSA), Alexa Fluor™ 647 conjugate

Catalog No. A34785
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A34785 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A34785 Supplier Invitrogen™ Supplier No. A34785
Only null left

Suited for applications in which well-defined molecular weight tracers are needed

  • Applications include studying the rate of endocytosis and exocytosis and following intracellular protein processing

Cell Analysis, Cell Tracing and Tracking, Cellular Imaging, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining and Detection

Order Info

Shipping Condition: Room temperature

Specifications

Quantity 5 mg
Conjugate Alexa Fluor 647
Species Bovine
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label Type Alexa Fluor Dyes
Shipping Condition Room Temperature
Form Lyophilized
Protein Subtype Albumin Proteins
Product Line Alexa Fluor

For Research Use Only. Not for use in diagnostic procedures.