Learn More
Research Products Corporation Water, Molecular Biology Grade, DNase and RNase Free, 10 Liter Cubitainer
Catalog No. 50476562
Encompass
Certified RNase and DNase free for use in molecular biology applications. Ideal for making reagents, rinsing glass and plastic ware, enzymatic reactions and more. Toxic agents such as DEPC are not used in purification procedures.
  • Endotoxin Free
  • RNase Free
  • DNase Free
*Tested to < 0.001 EU/ML
Meets or exceeds CAP/CLSI specifications for Clinical Laboratory Reagent Water (CLRW) and USP/NF Purified Water.
Catalog No. 50476562 Supplier Research Products Corporation Supplier No. RPI248796
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Certified RNase and DNase free for use in molecular biology applications. Ideal for making reagents, rinsing glass and plastic ware, enzymatic reactions and more. Toxic agents such as DEPC are not used in purification procedures.
  • Endotoxin Free
  • RNase Free
  • DNase Free
*Tested to < 0.001 EU/ML
Meets or exceeds CAP/CLSI specifications for Clinical Laboratory Reagent Water (CLRW) and USP/NF Purified Water.