Learn More
Moldex™ SparkPlugs™ PlugStation™ Ear Plug Dispenser
Catalog No. 19384353
Change view
Click to view available options
Corded:
Corded
Uncorded
Metal Detectable:
No
Yes
Holds:
150 Pairs
250 Pairs
500 Pairs
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Corded Metal Detectable Holds
19384353 Corded Yes 150 Pairs
19003410A Uncorded No 250 Pairs
19003410B Uncorded No 500 Pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19384353 Supplier Moldex™ Supplier No. 6881
SparkPlugs™ PlugStation™ dispensers are pre-filled with ear plugs and are convenient for any work environment. Easy access, wall-mountable unit will help ensure compliance. When the PlugStation is empty, simply dispose of it and hang a new one.
  • Holds SparkPlugs™ disposable ear plugs
  • PlugStations™ are sealed to ensure ear plugs stay clean and sanitary
  • High visibility to increase compliance
  • Dispenses one pair at a time to eliminate wasteful grabbing
  • Includes mounting bracket
Specifications
ANSI S3.1974 Yes
Cord Color Blue
Cord Material PVC-free
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Easy Access to Hearing Protection for Food Processing Workers and Facility Visitors in Reception Areas, Break Rooms, Workstations, and Other Noisy Environments.
High Visibility Yes
Holds 150 Pairs
Packaging Type Dispenser
Pairs per Package 150 Pairs/Dispenser
Plug Material Foam
Plug Color Blue, White
Noise Reduction Rating (NRR) 33 dB
Corded Corded
Plug Shape Bullet
Disposable Single-use
Metal Detectable Yes
Material PVC-free Plastic
Mounting Wall Mountable
Includes Mounting Hardware
Show More Show Less