missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Moldex™ 7000 Series Half-Mask P100 Pre-Assembled Respirators
Description
- P100 particulate filter pre-assembled respirator with Series 7000 facepiece
- Series sleek design for modern, high-tech look and increased worker acceptance
- Lightweight design plus extra-wide sealing area provides all-day comfort
- Low profile, compact design fits well under welding helmets and with safety glasses
- Mask drops down for convenient storage around the neck or locks down for custom fit
- Adjustable head cradle and curved neck buckles for extra comfort
- Wide field of vision
Specifications
Specifications
| Certifications (NIOSH) | NIOSH 42 CFR Part 84 |
| Connection Type | Bayonet |
| Color | Blue |
| Filter Type | P100 |
| Material | Thermoplastic Elastomer |
| Product Type | Pre-Assembled Half-Mask Respirator |
| Size | Small |
| For Use With (Application) | Degreasing, forging, foundries, heat treating operations, paper making, rendering, rubber manufacturing, pharmaceuticals, petrochemicals |
| For Use With (Equipment) | Series 8910 N95, Series 8970 R95 or Series 7940 P100 filters |
| Includes | P100 Particulate Filter Disks |
| Show More |