missing translation for 'onlineSavingsMsg'
Learn More

Moldex™ 7000 Series Half-Mask P100 Pre-Assembled Respirators

Catalog No. 19384336
Change view
Click to view available options
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19384336 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19384336 Supplier Moldex™ Supplier No. 7943

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Lightweight 7000 Series half-mask respirators are specifically designed to comfortably conform to facial contours and with a modern look. Pre-assembled with 7940 P100 filter disks.

  • P100 particulate filter pre-assembled respirator with Series 7000 facepiece
  • Series sleek design for modern, high-tech look and increased worker acceptance
  • Lightweight design plus extra-wide sealing area provides all-day comfort
  • Low profile, compact design fits well under welding helmets and with safety glasses
  • Mask drops down for convenient storage around the neck or locks down for custom fit
  • Adjustable head cradle and curved neck buckles for extra comfort
  • Wide field of vision

Specifications

Certifications (NIOSH) NIOSH 42 CFR Part 84
Connection Type Bayonet
Color Blue
Filter Type P100
Material Thermoplastic Elastomer
Product Type Pre-Assembled Half-Mask Respirator
Size Large
For Use With (Application) Degreasing, forging, foundries, heat treating operations, paper making, rendering, rubber manufacturing, pharmaceuticals, petrochemicals
For Use With (Equipment) Series 8910 N95, Series 8970 R95 or Series 7940 P100 filters
Includes P100 Particulate Filter Disks
Model 7943
Strap Attachment Type Drop-down, Lock-down
Show More Show Less