Learn More
Moldex™ Jetz™ Reusable Ear Plugs
Catalog No. 19384340
Change view
Click to view available options
Corded:
Corded
Catalog No. Corded
19384340 Corded
1 options
Catalog No. 19384340 Supplier Moldex™ Supplier No. MDX6455
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Jetz™ special grip at the plug stem makes insertion and removal simple, even when wearing gloves. Extra-soft material provides all-day comfort.
  • Extra-soft material and triple flange design seals gently without pressure
  • Plugs can be washed and reused
  • 100% PVC-Free™
Specifications
ANSI S3.1974 Yes
Cord Color Green
Cord Material Cloth
CSA Z94.2 Class BL
Size Universal
For Use With (Application) Construction, Utilities, Manufacturing, Production, Mining, Oil and Gas, and Other Various Industries
High Visibility Yes
Packaging Type Poly bag
Pairs per Package 50 Pairs/Pk.
Plug Material TPE
Plug Color Green
Noise Reduction Rating (NRR) 27 dB
Corded Corded
Plug Shape Flanged
Product Type Reusable Earplugs
Disposable Reusable
Metal Detectable No
Dielectric No
Show More Show Less