missing translation for 'onlineSavingsMsg'
Learn More

Moldex™ HandyStrap™ Series R95 Particulate Respirators

Catalog No. 19046758
Change view
Click to view available options
Color:
Gray
Size:
Medium/Large
With Exhalation Valve:
Yes
Nuisance Odor Relief:
Acid Gases
None
Ozone and Organic Vapors
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size With Exhalation Valve Nuisance Odor Relief
19046758 Gray Medium/Large Yes None
19046756 Gray Medium/Large Yes Ozone and Organic Vapors
19999367 Gray Medium/Large Yes Acid Gases
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19046758 Supplier Moldex™ Supplier No. 2740R95

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R95 disposable respirators feature a unique HandyStrap™ with buckle, allowing the mask to hang down around the user’s neck when not in use.

  • Cloth strap is cool and comfortable against the skin
  • HandyStrap™ with buckle, allows mask to hang down around the user’s neck when not in use preventing premature disposal
  • Dura-Mesh™ provides bi-directional protection from dirt on the outside and moisture on the inside
  • Soft foam nose cushion for added comfort and no pressure points
  • Softspun™ lining for increased comfort and durability
  • Ventex™ exhalation valve lets the hot air out faster to enable easier breathing
  • Meets heat and flame resistance in accordance with ANSI/ISEA 110-2003 Section 7.11.1
  • R95 Series Applications: Grinding, torch cutting, sanding, metal pouring, sweeping, metal working, bagging, foundries, stone quarrying, agriculture, metal machining, construction, cement, ceramics manufacturing, underground mining, timber/lumber mills, polishing, buffing, lime, poultry, textiles, welding, brazing, soldering
  • R95 Plus Nuisance Levels of Ozone and Organic Vapor Applications (Fisher cat #19-046-756 and 19-046-757 only): Welding, arc cutting, foundry operations, brazing, plasma spraying, laser beam cutting/welding, laboratory settings, agriculture, petrochemical manufacturing, rendering plants, auto repair, mining operations, waste disposal
  • R95 Plus Nuisance Levels of Acid Gases Applications (Fisher cat #19-999-367 and 19-999-368 only): Welding, arc cutting, foundry operations, brazing, plasma spraying, laser beam cutting/welding, laboratory settings, agriculture, petrochemical manufacturing, rendering plants, auto repair, mining operations, waste disposal

Specifications

Aerosol Type R - Somewhat Resistant to Oil
ANSI Standard ANSI/ISEA 110-2003, Section 7.11.1
Certifications (NIOSH) NIOSH 42 CFR 84, R95
Color Gray
Design Type Cup/Molded
Disposable Single-use
Filter Type R95
Material Dura-Mesh
Product Type General Purpose Particulate Respirator
Size Medium/Large
With Exhalation Valve Yes
Filter Efficiency 95%
For Use With (Application) Foundry operation, lab settings, petrochemical and chemical processing, printing, mortuary, sewage
Material Headstrap Cloth
Model 2740R95
Nose Piece Material Soft foam
Nuisance Odor Relief None
Quantity 10/Pk.
Strap Attachment Type HandyStrap™ with Buckle
FDA Cleared Surgical Mask No
Show More Show Less