Learn More
Moldex™ 7000 Series Reusable Half Mask Respirator Facepiece
Catalog No. p-4186928
Change view
Click to view available options
Format:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
19384318 Small
19384319 Medium
19384320 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19384318 Supplier Moldex™ Supplier No. MDX7001
Only null left
Sleek design for modern, high-tech look and increased worker acceptance
  • 100% PVC-free
  • Lightweight design plus extra-wide sealing area provides all-day comfort
  • Low profile, compact design fits well under welding helmets and with safety glasses
  • Mask drops down for convenient storage around the neck or locks down for custom fit
  • Adjustable head cradle and curved neck buckles for extra comfort
  • Wide field of vision
  • Simple maintenance with very few replacement parts
Compliance
Meets EN 140; CE requirements.
Specifications
Certifications/Compliance EN 140:1998, CE
Material Thermoplastic Elastomer, PVC
Product Type Half Mask Respirator Facepiece
Format Small
Includes Adjustable head cradle and curved neck buckle