Learn More
Moldex™ 7000 Series Reusable Half Mask Respirator Facepiece
Description
Sleek design for modern, high-tech look and increased worker acceptance
- 100% PVC-free
- Lightweight design plus extra-wide sealing area provides all-day comfort
- Low profile, compact design fits well under welding helmets and with safety glasses
- Mask drops down for convenient storage around the neck or locks down for custom fit
- Adjustable head cradle and curved neck buckles for extra comfort
- Wide field of vision
- Simple maintenance with very few replacement parts
Compliance
Meets EN 140; CE requirements.
Specifications
Specifications
| Certifications/Compliance | EN 140:1998, CE |
| Material | Thermoplastic Elastomer, PVC |
| Product Type | Half Mask Respirator Facepiece |
| Format | Large |
| Includes | Adjustable head cradle and curved neck buckle |